Electronic overhead stirrers
Supplier: VELP SCIENTIFIC
F201A0150 F201A0152 F201A0151 F201A0155 F201A0156 F201A0157 SA201A0155 SA201A0151
441-0345EA 1160 EUR
441-0345 441-0347 441-0346 441-0348 441-0349 441-0350 441-0413 441-0474
Electronic overhead stirrers
Stirrers Overhead Stirrers
Range of overhead stirrers with electronic adjustment that are equipped with advanced safety systems.
  • Compact, easy to use ergonomic design, units are lightweight, reliable and quiet
  • Progressive start-up prevents liquid from coming out of the vessel
  • Microprocessor controlled motor means constant speed, even when the viscosity of the liquid changes (except ES models)
  • Model BS, PW, DLS, LH and DLH have safety chuck without key, which helps ensure safe use
  • DLS and DLH models have two digital displays showing both actual and set speeds simultaneously
Order Now
SPECIFICATIONS
Model PW ES LS DLS LH DLH
Environmentally Preferable
Max. viscosity 100000 mPas 1000 mPas 25000 mPas 25000 mPas 50000 mPas 50000 mPas
Speed range 20 - 1200 min⁻¹ 50 - 1300 min⁻¹ 50 - 2000 min⁻¹ 50 - 2000 min⁻¹ 50 - 2000 min⁻¹ 50 - 2000 min⁻¹
Max. torque at chuck 120 Ncm 15 Ncm 40 Ncm 40 Ncm 80 Ncm 80 Ncm
W×D×H 80×196×230 mm 80×200×160 mm 80×196×215 mm 80×196×215 mm 80×196×230 mm 80×196×230 mm
Weight 2,9 kg 1,3 kg 2,3 kg 2,5 kg 2,9 kg 3,0 kg