2-Methyl-4-trimethylsilyl-1-buten-3-yne ≥97%

Supplier: Thermo Fisher Scientific
Warning

44189.04 44189.09 44189.18
44189.04EA 50.2 EUR
44189.04 44189.09 44189.18
2-Methyl-4-trimethylsilyl-1-buten-3-yne ≥97%
2-Methyl-4-trimethylsilyl-1-buten-3-yne

Purity: 97% CAS No.: 18387-60-5 Molecular Formula: (CH3)3SiC≡CC(CH3)=CH2 Formula Weight: 138.28 Physical Form: Liquid Density: 0.778 Boiling Point: 32°/15mm UN No.: UN1993 MDL No.: MFCD00190206

Formula: (CH₃)₃SiC ΞCC(CH₃)=CH₂
Boiling Pt: 32 °C (15 mmHg)
Density: 0,778
Storage Temperature: Ambient
MDL Number: MFCD00190206
CAS Number: 18387-60-5
ADR: 3,III

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR