Cycloheximide ≥90.0%

Supplier: VWR Chemicals
Danger

Synonyms: Naramycin A, Actidione, 3-[2-(3,5-Dimethyl-2-oxocyclohexyl)-2-hydroxyethyl]glutarimide,

441894CEA 751 EUR
441894C
Cycloheximide ≥90.0%
Cycloheximide

Inhibits DNA and protein synthesis.

Formula: C₁₅H₂₃NO₄
MW: 281.35 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00082346
CAS Number: 66-81-9
EINECS: 200-636-0
UN: 2811
ADR: 6.1 I
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3abdf7ad3f7b3ea9e67fba3b6f3970c6627794be&vid=6f07d0130a35bc66b22ecee29fc6c31b20823099&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay Min. 90.0 %
Melting point 107 - 120 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR