VWR®, Magnetic stirring bars, plain
Supplier: VWR Collection
442-0262EA 21.5 EUR
442-0262 442-0263 442-0264 442-0265 442-0266 442-0267 442-0268 442-0269 442-0270 442-0271 442-0272 442-0273 442-0274 442-0275 442-0276 442-0277 442-0278 442-0279 442-0280
VWR®, Magnetic stirring bars, plain
Stirrers Magnetic Stirring Bars
PTFE covered, strong Alnico V magnetic core. Smooth surface with rounded edges that intensify the stirring effect, even at low speeds.
  • Excellent chemical resistance
  • Temperature resistant, suitable for high and low temperature applications (−200 to +280 °C)