Magnetic stirrers, MIXdrive series
Supplier: 2MAG
40300 40301 40303 40201 40101 40001 40302
442-0796EA 293 EUR
442-0796 442-0797 442-0798 442-0795 442-0794 442-0793 TMAG40302
Magnetic stirrers, MIXdrive series
Stirrers Magnetic Stirrers
The hermetically sealed stainless steel housing is water, germ and dust-proof. Operating temperatures for the basic version up to 50 °C, for HT version up to 95 °C submerged in water baths, up to 200 °C in ovens. M4 thread is integrated in the stainless steel bottom plate (only for MIXdrive 1 XS basic version).
  • Maintenance- and wear-free inductive drive
  • 100% synchronised jerk-free mixing
  • Submersible in water and oil baths
  • Suitable for use in CO₂ incubators
IP protection class: IP 68
External control unit (must be ordered separately):
SoftStart forMore Product Information