Magnetic hotplate stirrers, ARE/AREX 5 series
Supplier: VELP SCIENTIFIC
F20500560 F20500570 SA20500570 SA20500580 F20500580 SB20500580 SC20500580
442-1335EA 348 EUR
442-1335 442-1336 442-1338 442-1339 442-1337 442-1340 442-1406
Magnetic hotplate stirrers, ARE/AREX 5 series
Stirrers Magnetic Hotplate Stirrers
Magnetic hotplate stirrers, designed to provide optimal heating performance, reliability, and exceptionally long product lifetime. Different models to fulfil any laboratory requirement, from basic applications to precise thermoregulation.
  • Excellent heating performance up to 310 °C with homogeneous temperature
  • Precise thermoregulation of the medium with PT100/PT1000 probe (±1,0 °C) and VTF digital thermoregulator (±1,0 °C)
  • Powerful brushless motor gives superior resistance over time and top-notch stirring performance, for up to 20 L (H₂O)
  • The internal
More Product Information