VWR®, Magnetic stirring bars, cylindrical
Supplier: VWR Collection
442-4520EA 23.3 EUR
442-4520 442-4521 442-4522 442-4523 442-4524 442-4525 442-4530 442-4527 442-4528 442-4529 442-0259 442-0260 442-0261 442-0295
VWR®, Magnetic stirring bars, cylindrical
Stirrers Magnetic Stirring Bars
PTFE covered, strong Alnico V magnetic core. Rounded, smooth surface.
  • Excellent chemical resistance
  • Temperature resistant, suitable for use at high and low temperatures (−200 to +280 °C)