Sodium dodecyl sulphate (SDS) 99,0 Biochemistry, purified_specially

Supplier: VWR Chemicals
Danger

Synonyms: Dodecyl sulphate sodium salt, Sodium dodecyl sulfate, Dodecyl sulphuric acid sodium salt, Lauryl sulphate sodium salt, Sodium lauryl sulphate

44215HNEA 456 EUR
44215HN 44215LT
Sodium dodecyl sulphate (SDS) 99,0 Biochemistry, purified_specially
Sodium dodecyl sulphate (SDS)
Formula: CH₃(CH₂)₁₁OSO₃Na
MW: 288.38 g/mol
Melting Pt: 204…207 °C
Density: 1.1 g/cm³ (20 °C)
Flash Pt: 100 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00036175
CAS Number: 151-21-3
EINECS: 205-788-1
UN: 1325
ADR: 4.1 III
REACH: 01-2119489461-32
Merck Index: 14,08636
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fe4cb1741fba41f699a3be006d0b01b1b551898c&vid=c6df9a86fc351b319e68cc79c1c17f733b4fec67&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (calculated on anhydrous) Min. 99.0 %
Assay (C12 compounds)(calc.on anhydrous) Min. 98.5 %
IR Spectrum Passes test
Cl (Chloride) Max. 0.03 %
PO4 (Phosphate) Max. 1 ppm
Pb (Lead) Max. 2 ppm
Absorbance (220-350 nm) (3 %) Max. 0.1


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR