Octakis(trimethylsiloxy)silsesquioxane

Supplier: Thermo Fisher Scientific
Warning

44231.14
44231.14EA 0 EUR
44231.14
Octakis(trimethylsiloxy)silsesquioxane
Octakis(trimethylsiloxy)silsesquioxane

CAS No.: 51777-38-9 Molecular Formula: C24H72O20Si16 Formula Weight: 1130.18 Physical Form: Powder Appearance: White MDL No.: MFCD01310212

Formula: C₂₄H₇₂O₂₀Si₁₆
Storage Temperature: Ambient
MDL Number: MFCD01310212
CAS Number: 51777-38-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR