Zinc oxide ≥99.9% (metals basis)

Supplier: Thermo Fisher Scientific
Warning

44263.30 44263.A1 44263.A7
44263.30EA 50.5 EUR
44263.30 44263.A1 44263.A7
Zinc oxide ≥99.9% (metals basis)
Zinc oxide

Purity: 99.9% CAS No.: 1314-13-2 Molecular Formula: ZnO Formula Weight: 81.37 Physical Form: -200 Mesh Powder Density: 5.606 Melting Point: 1975° Merck Reference: 14,10147 UN No.: 3077 MDL No.: MFCD00011300

Formula: ZnO
Melting Pt: 1975 °C
Density: 5,606
Storage Temperature: Ambient
MDL Number: MFCD00011300
CAS Number: 1314-13-2
EINECS: 215-222-5
UN: 3077
ADR: 9,III
REACH: 01-2119463881-32
Merck Index: 14,10147

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR