Aluminium ≥99.9999% (metals basis), rods, Puratronic®, Ø 22 mm (0.87 in)

Supplier: Thermo Fisher Scientific

44336.BQ
44336.BQEA 0 EUR
44336.BQ
Aluminium ≥99.9999% (metals basis), rods, Puratronic®, Ø 22 mm (0.87 in)
Aluminium

Purity: 99.9999% CAS No.: 7429-90-5 Physical Form: ≈10.35g/cm Odour: Odourless MDL No.: MFCD00134029

Formula: Al
Boiling Pt: 2327 °C (1013 hPa)
Melting Pt: 660 °C
Density: 2,7 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134029
CAS Number: 7429-90-5
EINECS: 231-072-3
Merck Index: 13,00321

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR