Incubating shakers, MaxQ™ 6000
Supplier: Thermo Fisher Scientific
SHKE6000-1CE SHKE6000-8CE SHKE6000-8CEPR SHKE6000-1CEPR
444-0650EA 14200 EUR
444-0650 444-0651 444-1414 444-1070
Incubating shakers, MaxQ™ 6000
Shakers Shaking Incubators
Digital high capacity incubating shakers which combine orbital shaking with general purpose incubation for added versatility. Both refrigerated and standard units have two adjustable-height shelves which provide added storage and the door can be hinged from either the left or right by the user for convenient placement in the lab. The spacious stainless steel chamber allows stacking of a 457×457 mm platform to double the shaking capacity. Applications include bacterial suspensions, solubility studies, extraction procedures, staining and destaining, hybridisations, diagnostic tests, washingMore Product Information