Bead mixers, Bead Genie™
Supplier: SCIENTIFIC INDUSTRIES ELMIS
SI-B103
444-1428EA 0 EUR
444-1428
Bead mixers, Bead Genie™
Homogenisers Homogenisers Bench
Bead Genie™ is ideal for bead beating, lysing, extraction, or homogenisation applications.
  • Works with microtubes (snap cap or screw cap), 6×15 ml tubes, or 3×50 ml tubes
  • Supplied with tube holders
  • No tools required to set up or operate
  • Programmable timer or continuous 'RUN' settings
  • Variable 'RUN/STOP' timer for superior interval mixing
  • Two year parts and labour warranty
It is ideal for lysing, extractions, or homogenisation applications, utilizing an aggressive vertical mixing motion. Unique vertical motion ensures fast and efficient processing ofMore Product Information