Cerium(III) carbonate hydrate ≥99.999% (REO, rare earth oxide basis), REacton®

Supplier: Thermo Fisher Scientific
Warning

44496.14 44496.22 44496.36
44496.14EA 122 EUR
44496.14 44496.22 44496.36
Cerium(III) carbonate hydrate ≥99.999% (REO, rare earth oxide basis), REacton®
Cerium (III) carbonate hydrate

Purity: 99.999% CAS No.: 54451-25-1 Molecular Formula: Ce2(CO3)3.xH2O Formula Weight: 460.27(anhy) Physical Form: Crystalline Melting Point: 848° MDL No.: MFCD00149635

Formula: C₃Ce₂O₉·H₂O
Melting Pt: 848 °C
Storage Temperature: Ambient
MDL Number: MFCD00149635
CAS Number: 54451-25-1
EINECS: 208-655-6
Merck Index: 14,1996

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR