Calcium-D gluconate 2,5% (w/w) in aqueous solution, gel

Supplier: Thermo Fisher Scientific

Synonyms: Calcium Gluconate, Calcium (2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate, Gluconate Calcium, D-Gluconic acid calcium salt

44542.15 44542.TJ
44542.15EA 160 EUR
44542.15 44542.TJ
Calcium-D gluconate 2,5% (w/w) in aqueous solution, gel
Calcium D-gluconate

CAS No.: 299-28-5 Molecular Formula: Ca[HOCH2(CHOH)4CO2]2 Formula Weight: 430.37 Physical Form: Gel MDL No.: MFCD00064209

Formula: HOH₂C(CH(OH))₄COOCa
Storage Temperature: Ambient
MDL Number: MFCD00064209
CAS Number: 299-28-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR