Trifluoroacetic acid ≥99.5% for HPLC

Supplier: Thermo Fisher Scientific
Danger

44630.AP 44630.AY
44630.APEA 531 EUR
44630.AP 44630.AY
Trifluoroacetic acid ≥99.5% for HPLC
Trifluoroacetic acid

Purity: 99.5+% CAS No.: 76-05-1 Molecular Formula: CF3CO2H Formula Weight: 114.02 Physical Form: Liquid Density: 1.480 Refractive Index: 1.2840 Sensitivity: Hygroscopic Odour: Pungent Melting Point: -15° Boiling Point: 72-73° Beilstein Reference: 742035 Merck Reference: 14,9681 UN No.: 2699 MDL No.: MFCD00004169

Formula: CF₃COOH
Boiling Pt: 72…73 °C
Melting Pt: –15 °C
Density: 1,480
Storage Temperature: Ambient
MDL Number: MFCD00004169
CAS Number: 76-05-1
EINECS: 200-929-3
UN: 2699
ADR: 8,I
Merck Index: 14,9681
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9501f3ac85d66e8d19d1cd1c3fe5409b974e9d8a&vid=be6aa627129b49ccda76006e7003b46d79b933cc&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR