Nickel ≥99.5% (metals basis), foil, Thickness: 1.0 mm (0.04 in)

Supplier: Thermo Fisher Scientific
Danger

44824.FI 44824.GH 44824.HP
44824.FIEA 50.6 EUR
44824.FI 44824.GH 44824.HP
Nickel ≥99.5% (metals basis), foil, Thickness: 1.0 mm (0.04 in)
Nickel

Purity: 99.5% CAS No.: 7440-02-0 Physical Form: ≈22.3g/50x50mm Odour: Odourless MDL No.: MFCD00011137

Formula: Ni
Boiling Pt: 3007 °C (1013 hPa)
Melting Pt: 1453 °C
Density: 8,9 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011137
CAS Number: 7440-02-0
EINECS: 231-111-4
UN: 3089
ADR: 4.1,III
Merck Index: 12,06582

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR