Sulphuric acid solution_standardised 0.05 N aqueous solution

Supplier: Thermo Fisher Scientific

45048.K2
45048.K2EA 37.1 EUR
45048.K2
Sulphuric acid solution_standardised 0.05 N aqueous solution
Sulphuric acid

Molecular Formula: H2SO4 Formula Weight: 98.08 Physical Form: Liquid Odour: Odourless MDL No.: MFCD00064589

Formula: H₂SO₄
MW: 98.08 g/mol
Boiling Pt: 330
Melting Pt: 10.38
Density: 1.01 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064589
CAS Number: 7664-93-9
EINECS: 231-639-5
UN: 2796
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR