4-(N,N-Dimethyl)aniline magnesium bromide 0.5 M in THF, AcroSeal®

Supplier: Thermo Fisher Scientific
Danger

450610500
450610500.EA 199 EUR
450610500.
4-(N,N-Dimethyl)aniline magnesium bromide 0.5 M in THF, AcroSeal®
4-(N,N-Dimethyl)aniline magnesium bromide
Formula: C₈H₁₀BrMgN
Density: 0,951 g/cm³
Flash Pt: –17 °C
Storage Temperature: Ambient
MDL Number: MFCD00672006
CAS Number: 7353-91-5
UN: 2924
ADR: 3,II

Order Now

Specification Test Results

Appearance Yellow to brown Liquid
Titration Grignard 0.40 to 0.60 M


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR