Gold ≥99.999% (metals basis), wire, Premion®, Ø 0.35 mm (0.0138 in)

Supplier: Thermo Fisher Scientific

45084.CC 45084.BY 45084.BU
45084.CCEA 2710 EUR
45084.CC 45084.BY 45084.BU
Gold ≥99.999% (metals basis), wire, Premion®, Ø 0.35 mm (0.0138 in)
Gold

Purity: 99.999% CAS No.: 7440-57-5 Physical Form: ≈1.82g/m Odour: Odourless MDL No.: MFCD00003436

Formula: Au
Boiling Pt: 2800 °C (1013 hPa)
Melting Pt: 1063 °C
Density: 19,3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003436
CAS Number: 7440-57-5
EINECS: 231-165-9
Merck Index: 12,04526

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR