Antimony ≥99% (metals basis), shot ≤ 6.35 mm (0.25 in)

Supplier: Thermo Fisher Scientific
Warning

45095.22 45095.A1 45095.36
45095.22EA 52.3 EUR
45095.22 45095.A1 45095.36
Antimony ≥99% (metals basis), shot ≤ 6.35 mm (0.25 in)
Antimony

Purity: 99% CAS No.: 7440-36-0 Odour: Odourless MDL No.: MFCD00134030

Formula: Sb
Boiling Pt: 1637 °C (1013 hPa)
Melting Pt: 630 °C
Density: 6,684 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134030
CAS Number: 7440-36-0
EINECS: 231-146-5
UN: 2871
ADR: 6.1,III
Merck Index: 12,00733

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR