Fuego SCS basic
Supplier: WARTEWIG
8.201.000
451-0114EA 526.27 EUR
451-0114
Fuego SCS basic
Heaters Gas Burners
This model of Fuego series sets new standards in laboratory gas burner technology for more safety with a stainless steel foot pedal and button function. Madeup of stainless steel and with fireproof controls.
  • Simply safe to operate, SCS safety control system
  • Three different programs are available
  • Flaming with button function
  • Easy to clean - a laboratory gas burner without slots, corners and edges
  • Ignition and flame monitoring and overtemperature protection
  • Excellent energy values ​​reduce gas consumption, passive safety
Suitable for all flame-relatedMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable