Benzamidoxime 97%

Supplier: Thermo Fisher Scientific
Danger

453280010 453280050
453280010.EA 37 EUR
453280010. 453280050.
Benzamidoxime 97%
Benzamidoxime
Formula: C₇H₈N₂O
Melting Pt: 74…77 °C
Storage Temperature: Ambient
MDL Number: MFCD00031485
CAS Number: 613-92-3
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to off-white Solid
Infrared spectrum Conforms
Melting point 73 to 79 °C
HPLC ≥94.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR