Supelco® PESTANAL® Organic Reference Standard, Desmedipham

Supplier: Merck

45426-50MG
45426-50MGEA 74.5 EUR
45426-50MG
Supelco® PESTANAL® Organic Reference Standard, Desmedipham
Standards Organic Standards

  • GHS signal: GHS09


Caution: P202 - P273 - P280 - P308 + P313 - P391 - P405

https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=54bea2bab0a2060e778915246858153a4d63ca8c&vid=54bea2bab0a2060e778915246858153a4d63ca8c&prid=web&clid=SAPDAM

Order Now

SPECIFICATIONS

Environmentally Preferable
M-Clarity quality level MQ100
CAS No. 13684-56-5
MDL No. MFCD00055462
Formula C₁₆H₁₆N₂O₄
Molecular weight 300,31
Hazard class description H361d - H410
Flash point Not applicable
Shipping conditions STD
Storage conditions 2...30 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR