Anti-Pan-NMDA NR1 (R1JHL) Mouse Monoclonal Antibody [clone: R1JHL]

Supplier: Merck Millipore (Oncogene)
454578-15
454578-15EA 796 EUR
454578-15
Anti-Pan-NMDA NR1 (R1JHL) Mouse Monoclonal Antibody [clone: R1JHL]
Antibodies
Type: Primary
Antigen: Pan-NMDA NR1 (R1JHL)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR