Fluorocyclohexane 97%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Cyclohexyl fluoride

459320010
459320010.EA 233 EUR
459320010.
Fluorocyclohexane 97%
Fluorocyclohexane
Formula: C₆H₁₁F
Boiling Pt: 103…105 °C
Melting Pt: 13 °C
Density: 0,928 g/cm³
Flash Pt: 5 °C
Storage Temperature: Ambient
MDL Number: MFCD00021285
CAS Number: 372-46-3
UN: 1993
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4140 to 1.4160 (20°C, 589 nm)
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR