Bis[di-tert-butyl(4-dimethylaminophenyl)phosphine]palladium(0) (16,7% Pd)

Supplier: Thermo Fisher Scientific
Warning

46000.MD 46000.03
46000.MDEA 132 EUR
46000.MD 46000.03
Bis[di-tert-butyl(4-dimethylaminophenyl)phosphine]palladium(0) (16,7% Pd)
Bis[di-tert-butyl(4-dimethylaminophenyl)phosphine]palladium(0)

CAS No.: 1233717-68-4 Molecular Formula: C32H56N2P2Pd Formula Weight: 637.18 Physical Form: Crystalline Sensitivity: Air Sensitive Appearance: Off-white MDL No.: MFCD15071402

Formula: C₃₂H₅₆N₂P₂Pd
Storage Temperature: Ambient
MDL Number: MFCD15071402
CAS Number: 1233717-68-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR