Aluminium oxide, Drysphere®, without indicator

Supplier: Thermo Fisher Scientific

Synonyms: Aluminium(III) oxide, Alumina C, Aluminum oxide

46093.A1 46093.30
46093.A1EA 119 EUR
46093.A1 46093.30
Aluminium oxide, Drysphere®, without indicator
Aluminium oxide

CAS No.: 1344-28-1 Molecular Formula: Al2O3 Formula Weight: 101.96 Physical Form: 3.175mm Spheres Refractive Index: 1.765 Appearance: White Odour: Odourless Melting Point: 2045° Boiling Point: 2980° MDL No.: MFCD00003424

Formula: Al₂O₃
Boiling Pt: 2980 °C
Melting Pt: 2045 °C
Storage Temperature: Ambient
MDL Number: MFCD00003424
CAS Number: 1344-28-1
EINECS: 215-691-6
Merck Index: 14,356

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR