Cooling circulators, with natural refrigerant
Supplier: HUBER
2009.0002.01 2010.0002.01 2008.0005.01
461-0142EA 7360 EUR
461-0142 461-0143 461-0564
Cooling circulators, with natural refrigerant
Circulating Thermostats Open Circulator Systems Baths with Detachable Circulators
These circulators, with insulated refrigeration baths, are designed for direct temperature control from −30 up to +200 °C.
  • Pilot ONE® is interchangeable and can be used as a remote control (with an optional data cable)
  • Controller can be upgraded with enhanced functionality at any time (Exclusive and Professional E-grade® packages), basic version features six languages (English, German, French, Spanish, Italian and Russian)
  • The display resolution is 0,1 °C; display can be customised to the users' preferences and important information e.g. set point,
More Product Information