Palladium 1% on carbon, granules, reduced

Supplier: Thermo Fisher Scientific

46462.14 46462.22 46462.36
46462.14EA 153 EUR
46462.14 46462.22 46462.36
Palladium 1% on carbon, granules, reduced
Palladium

CAS No.: 7440-05-3 Physical Form: 3mm (0.13in) Pellets MDL No.: MFCD00011167

Formula: Pd
MW: 106.420 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03457879
CAS Number: 7440-05-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR