Hydrochloric acid concentrate for dilution to 0.5 mol aqueous solution, DILUT-IT®, Laboratory Reagent, J.T.Baker®

Supplier: Avantor
Danger

Synonyms: Hydrochloric acid solution, Muriatic acid

4654.EA 36.88 EUR
4654.
Hydrochloric acid concentrate for dilution to 0.5 mol aqueous solution, DILUT-IT®, Laboratory Reagent, J.T.Baker®
Hydrochloric acid
Formula: HCl
MW: 36.46 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8 II

Order Now

Specification Test Results

Titer (mol/l) 0.4990-0.5010


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR