Ovens, OP series
Supplier: LTE SCIENTIFIC
OP60-U OP100-U OP150-U OP250-U OP60-UF OP100-UF OP150-UF OP250-UF OP60-M OP100-M OP150-M OP250-M OP60-MF OP100-MF OP150-MF OP250-MF
466-0104EA 2750 EUR
466-0104 466-0088 466-0092 466-0096 466-0105 466-0089 466-0093 466-0097 466-0102 466-0086 466-0090 466-0094 466-0103 466-0087 466-0091 466-0095
Ovens, OP series
Ovens Standard Ovens
Range of ovens with choice of either natural convection or forced convection by means of a fan. U (Uni-program) models offer single temperature selection/control at the push of a button. Following a mains power failure, the controller will automatically re-start. M (Multi-program) models will allow up to eight multiple-step cycles to be stored in the memory. Selectable temperature ramping function allows controlled temperature rates to be programmed into a cycle, repeated for a specified number of times or held continuously until interrupted. Following a mains power failure, recovery optionsMore Product Information