Drying and heating chambers with forced convection, FP series Avantgarde.Line
Supplier: Binder
9010-0363 9010-0366 9010-0365 9010-0364
466-0653EA 3780 EUR
466-0653 466-0656 466-0655 466-0654
Drying and heating chambers with forced convection, FP series Avantgarde.Line
Ovens Standard Ovens
The Binder FP series of material test chambers with mechanical convection and program functions provide reliably short drying times and particularly fast heating – even for chambers under full loads.
  • High temperature accuracy thanks to APT.line™ technology
  • Controller with interval and real-time programming
  • Adjustable fan speed
  • Electromechanical control of the exhaust air flap
  • Class 2 independent adjustable temperature safety device (DIN 12880) and class 3.1 independent temperature safety device (DIN 12880) with visual/acoustic alarm available for selection
More Product Information