Ovens, E 28
Supplier: Binder
9010-0001
466-3161EA 1290 EUR
466-3161
Ovens, E 28
Ovens Standard Ovens
Robust space-saving oven/steriliser with hydraulic mechanical thermostat and temperature setting via analogue dial indicator thermometer.
  • Adjustable ventilation flap
  • Option of models with or without Class 1 (DIN 12880) safety device
  • Timer 0 to 120 min
  • Analogue temperature display
IP protection class according to DIN EN 60529: IP 20
Certifications: IP protection class according to DIN EN 60529: IP 20
Delivery information: Supplied with two chrome plated shelves.