Drying ovens with natural convection, ED series
Supplier: Binder
9010-0075 9010-0167 9010-0168 9010-0169 9010-0190
466-3267EA 5960 EUR
466-3267 BIND9010-0167 BIND9010-0168 BIND9010-0169 466-3251
Drying ovens with natural convection, ED series
Ovens Standard Ovens
Universal drying ovens with stainless steel chambers which are ideal for drying and sterilisation applications in the pharmaceutical and chemical industries, biotechnology, medicine and universities. Ovens feature one temperature program ramp, an adjustable temperature safety device, Class 2 (DIN 12880) with visual alarm and adjustable ventilation by means of front ventilation slide and rear 50 mm Ø exhaust duct. GLP/GMP compliance is assisted by the optional RS422 interface and APT-COM™ DataControlSystem software.
  • APT.line™ preheating chamber technology with natural convection
  • DS
More Product Information