Indium ≥99.998% (metals basis), wire, Puratronic®, annealed, Ø 1.0 mm (0.04 in)

Supplier: Thermo Fisher Scientific

46661.G1 46661.G5 46661.G9
46661.G1EA 149 EUR
46661.G1 46661.G5 46661.G9
Indium ≥99.998% (metals basis), wire, Puratronic®, annealed, Ø 1.0 mm (0.04 in)
Indium
Formula: In
Boiling Pt: 2000 °C (1013 hPa)
Melting Pt: 156 °C
Density: 7,3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134048
CAS Number: 7440-74-6
EINECS: 231-180-0
Merck Index: 13,04971

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR