Supelco® VETRANAL® Organic Reference Standard, Progesterone

Supplier: Merck

46665-250MG
46665-250MGEA 359 EUR
46665-250MG
Supelco® VETRANAL® Organic Reference Standard, Progesterone
Standards Organic Standards

  • GHS signal: GHS08


Caution: P202 - P260 - P263 - P264 - P270 - P308 + P313

https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c955b80e2bea31de3c0dc41a4fab3079fa14a8ed&vid=c955b80e2bea31de3c0dc41a4fab3079fa14a8ed&prid=web&clid=SAPDAM

Order Now

SPECIFICATIONS

Synonym(s) 4-Pregnene-3,20-dione
Environmentally Preferable
M-Clarity quality level MQ100
CAS No. 57-83-0
MDL No. MFCD00003658
Molecular weight 314,46
Melting point 128 - 132 °C (lit.)
Hazard class description H351 - H360FD - H362
Shipping conditions STD
Storage conditions 2...30 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR