1,4-Benzenedimethanethiol

Supplier: Thermo Fisher Scientific
Warning

467610010 467610050
467610010.EA 119 EUR
467610010. 467610050.
1,4-Benzenedimethanethiol
1,4-Benzenedimethanethiol
Formula: C₈H₁₀S₂
Boiling Pt: 156°C (16hPa)
Storage Temperature: Ambient
MDL Number: MFCD00004872
CAS Number: 105-09-9
EINECS: 203-269-4
UN: 3335
ADR: 9,III

Order Now

Specification Test Results

Appearance white powder, crystals or solid
Infrared spectrum Conforms
GC ≥97.5 %
Melting point 40°C to 45°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR