Polyoxyethylene bis(amine), M.W. 1,000

Supplier: Thermo Fisher Scientific

Synonyms: PEG-bis-amine, Amine-PEG-Amine, O,O′-Bis(2-aminoethyl)polyethylene glycol, PEG-diamine, Amine-PEG-amine (MW 4000), Poly(ethylene glycol) diamine, Polyoxyethylene bis(amine), Diaminopolyethylene glycol

46833.MD 46833.03 46833.06
46833.MDEA 58.6 EUR
46833.MD 46833.03 46833.06
Polyoxyethylene bis(amine), M.W. 1,000
Polyoxyethylene bis(amine)

CAS No.: 24991-53-5 Molecular Formula: H2NCH2CH2(OCH2CH2)nNH2 Physical Form: Granular solid Sensitivity: Air Sensitive Appearance: White to off-white MDL No.: MFCD00132004

Formula: H₂NCH₂CH₂(OCH₂CH₂)nNH₂
Storage Temperature: Ambient
MDL Number: MFCD00132004
CAS Number: 24991-53-5

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR