Rebamipide

Supplier: Thermo Fisher Scientific
Danger

468590010 468590050 468590250
468590010.EA 56.3 EUR
468590010. 468590050. 468590250.
Rebamipide
Rebamipide
Formula: C₁₉H₁₅ClN₂O₄
Storage Temperature: Ambient
MDL Number: MFCD11114396
CAS Number: 90098-04-7
UN: 2811
ADR: 6.1,III
Merck Index: 13,08209

Order Now

Specification Test Results

Appearance white to beige crystalline powder
Infrared spectrum Conforms
HPLC ≥98.5 %
Loss on drying ≤2 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR