Dabigatran Etexilate 98%

Supplier: Thermo Fisher Scientific

469930010 469932500
469930010.EA 302 EUR
469930010. 469932500.
Dabigatran Etexilate 98%
Dabigatran Etexilate
Formula: C₃₄H₄₁N₇O₅
Boiling Pt: 827°C
Storage Temperature: Freezer
MDL Number: MFCD22422841
CAS Number: 211915-06-9

Order Now

Specification Test Results

Appearance white to beige powder
Infrared spectrum Conforms
HPLC ≥97.5 %
Melting point 128°C to 133°C
Water ≤5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR