Professional upright freezers, GGPv
Supplier: Liebherr
GGPV1470 GGPV6570 GGPV6590 GGPV1490 GGPV1470/924
472-1054EA 0 EUR
472-1054 472-1058 471-0916 471-0915 LIEBGGPV1470/924
Professional upright freezers, GGPv
Freezers Laboratory / General Purpose Freezers
Commercial freezer models featuring automatic defrost.
  • Indirect forced air cooling
  • Electronic temperature controller with digital display, temperature and alarms
  • Insulation 83 mm (temperature guaranteed at 43 °C)
  • Automatic defrosting with evaporation of condensates
  • Doors remain open at 90° and are self closing at less that 60°; reversible and lockable
Temperature range: from −10 to −26 °C.
Delivery information: These freezer cabinets are delivered with adjustable feet (150 to 180 mm), except models GGPV6570 and GGPV1470 which are supplied withMore Product Information