Ultra-low temperature freezers, upright, BioUltra UL570
Supplier: GRAM
UL570CG UL570LG
471-1111EA 11870 EUR
471-1111 471-1112
Ultra-low temperature freezers, upright, BioUltra UL570
Freezers Ultra Low Temperature Freezers
The BioUltra UL570 is an ultra-low temperature freezer that operates safely at −86 °C. By utilising high capacity components, cascade refrigeration system, latest generation vacuum insulated panel technology and insulated inner doors, the BioUltra cabinets are able to maintain safe and stable interior conditions. The BioUltra UL570 holds up to 384×50 mm boxes (using 4×4 racks), 192×75 mm boxes (using 3×4 racks) or 192×100 mm boxes (using 2×4 racks).
  • Cascade refrigeration system ensures high efficiency, performance and durability; easy access to heat
More Product Information