Blood Bank Refrigerators, PHCbi
Supplier: PHC Corporation
MBR-107D MBR-305GR MBR-705GR MBR-305GR-PE
MBR-107DEA 0 EUR
MBR-107D 471-1165 471-1166 SANYMBR-305GR-PE
Blood Bank Refrigerators, PHCbi
Refrigerators
With stainless steel interior and scratch resistant easy-to-clean baked-on acrylic finish. MBR series refrigerators provide two styles of storage systems, G-type (coated hard steel wire shelves) and GR-type (stainless steel roll-out drawers with card holders). A built-in controller with low and high temperature alarm maintains temperature at 2 °C (or 6 °C in case of malfunction) whilst a microprocessor is employed to maintain temperature within 1 °C of 4 °C, regardless of ambient temperature fluctuations and defrosting time.
  • Equipped with automatic 30-day recorder (MBR-107D/506D) or 1/7
More Product Information