Controlled-rate freezers, CryoMed
Supplier: Thermo Fisher Scientific
TSCM17XV TSCM34XV TSCM48XV TSCM17PV TSCM34PV TSCM48PV
471-1248EA 20040 EUR
471-1248 471-1249 471-1250 471-1313 471-1314 471-1359
Controlled-rate freezers, CryoMed
Freezers Cryogenic Freezers
In order to maintain cellular viability during the freezing process, certain cells such as protist, mammalian, and plant cells require precise control of the freezing rate in order to minimize the detrimental effects of undercooling and the heat liberated during the phase change process from water to ice. The Thermo Scientific™ CryoMed™ Controlled-Rate Freezers (CRFs) are designed with reliability, protection, and data traceability in mind for sample preparation and GMP production needs.
  • Intuitive touchscreen display allows easy set-up, operation, and
More Product Information