Controlled-rate freezers, CryoMed
Supplier: Thermo Fisher Scientific
TSCM17XV TSCM34XV TSCM48XV TSCM17PV TSCM34PV TSCM48PV
471-1248EA 20040 EUR
471-1248 471-1249 471-1250 471-1313 471-1314 471-1359
Controlled-rate freezers, CryoMed
Freezers Cryogenic Freezers
In order to maintain cellular viability during the freezing process, certain cells such as protist, mammalian, and plant cells require precise control of the freezing rate in order to minimize the detrimental effects of undercooling and the heat liberated during the phase change process from water to ice. The Thermo Scientific™ CryoMed™ Controlled-Rate Freezers (CRFs) are designed with reliability, protection, and data traceability in mind for sample preparation and GMP production needs.
  • Intuitive touchscreen display allows easy set-up, operation, and
More Product Information
Limited Time Sale
Select items below are currently on sale
Order Now
SPECIFICATIONS
Model CryoMed CRF 17 CryoMed CRF 34 CryoMed CRF 48 CryoMed CRF 17
with thermal printer
CryoMed CRF 34
with thermal printer
CryoMed CRF 48
Environmentally Preferable
Display Touch-screen LCD Touch-screen LCD Touch-screen LCD Touch-screen LCD Touch-screen LCD Touch-screen LCD
Alarm Audible and visual Audible and visual Audible and visual Audible and visual Audible and visual Audible and visual
Electrical 220 V, 50 - 60 Hz / 7FLA 220 V, 50 - 60 Hz / 7FLA 220 V, 50 - 60 Hz / 7FLA 220 V, 50 - 60 Hz / 7FLA 220 V, 50 - 60 Hz / 7FLA 220 V, 50 - 60 Hz / 7FLA
Door type Solid Solid Solid Solid Solid Solid
Door lock via access code via access code via access code via access code via access code via access code
Weight 69,8 kg 78,9 kg 86,6 kg 70,3 kg 79,3 kg 87,1 kg