General purpose upright freezers, TSG series
Supplier: Thermo Scientific
TSG400FESBW TSG400FESCW TSG700FESBW TSG700FESCW TSG1500FESBW TSG1500FESCW
471-1327EA 3374 EUR
471-1327 471-1329 471-1332 471-1334 471-1321 471-1323
General purpose upright freezers, TSG series
Freezers Laboratory / General Purpose Freezers
Designed for everyday laboratory needs the Thermo Scientific™ TSG series general purpose laboratory freezers are designed with your needs in mind. The TSG series range of freezers feature the performance, security, and quality required for your laboratory applications.
  • Freezers: −20 °C set point, settable range −25 to −10 °C
  • Wide range of sizes from solid door 400 to 1500 L double door
  • Whisper-quiet operation of <60 dBA
  • Adaptive defrost
  • White painted interior and exterior
  • Temperature, door ajar, and power failure (with battery backup) alarms
  • Low heat
More Product Information