Cadmium(II) bromide, ultra dry ≥99.999% (metals basis)

Supplier: Thermo Fisher Scientific
Warning


ampouled under argon

47105.14
47105.14EA 0 EUR
47105.14
Cadmium(II) bromide, ultra dry ≥99.999% (metals basis)
Cadmium (II) bromide

Purity: 99.999% CAS No.: 7789-42-6 Molecular Formula: CdBr2 Formula Weight: 272.22 Physical Form: Powder, Ampouled under argon Density: 5.192 Sensitivity: Hygroscopic Appearance: White to pale yellow Odour: Odourless Melting Point: 567° Boiling Point: 863° UN No.: UN2570 MDL No.: MFCD00015996

Formula: Br₂Cd
Boiling Pt: 863 °C
Melting Pt: 567 °C
Density: 5,192
Storage Temperature: Ambient
MDL Number: MFCD00015996
CAS Number: 7789-42-6
EINECS: 232-165-1
UN: 2570
ADR: 6.1,III
Merck Index: 14,1615

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR