Chromium(II) chloride ≥99.9% (metals basis) -80 mesh

Supplier: Thermo Fisher Scientific
Danger

47176.03 47176.06 47176.14
47176.03EA 63.2 EUR
47176.03 47176.06 47176.14
Chromium(II) chloride ≥99.9% (metals basis) -80 mesh
Chromium (II) chloride

Purity: 99.9% CAS No.: 10049-05-5 Molecular Formula: Cl2Cr Formula Weight: 122.9 Physical Form: -80 Mesh Powder Density: 2.878 Sensitivity: Air and Moisture Sensitive Appearance: Grey Odour: Odourless Melting Point: 824° UN No.: UN3260 MDL No.: MFCD00010947

Formula: Cl₂Cr
Melting Pt: 824 °C
Density: 2,878
Storage Temperature: Ambient
MDL Number: MFCD00010947
CAS Number: 10049-05-5
EINECS: 233-163-3
UN: 3260
ADR: 8,III
Merck Index: 14,2244

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR