Lenvatinib mesylate 98%

Supplier: Thermo Fisher Scientific

474520010 474522500
474520010.EA 488 EUR
474520010. 474522500.
Lenvatinib mesylate 98%
Lenvatinib mesylate
Formula: C₂₂H₂₃ClN₄O₇S
Storage Temperature: Refrigerator
MDL Number: MFCD18633219
CAS Number: 857890-39-2
New Product

Order Now

Specification Test Results

Appearance white to beige powder
Infrared spectrum Conforms
HPLC ≥97.5 %

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR